Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl Methacrylate 98.0+%, TCI America™
SDP

Catalog No. S08125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08125G 5 g
S081225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08125G Supplier TCI America Supplier No. S08125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38862-25-8
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.16
MDL Number MFCD15072193
InChI Key ACGJEMXWUYWELU-UHFFFAOYSA-N
Synonym Methacrylic Acid N-Succinimidyl Ester, N-Methacryloyloxysuccinimide
PubChem CID 10130244
IUPAC Name 2,5-dioxopyrrolidin-1-yl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 104°C
Color White
Quantity 5 g
Formula Weight 183.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl Methacrylate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.