accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CONNH

CAS RN 38869-47-5

IUPAC Name: 1-(4-methoxyphenyl)piperazine hydrochloridyl
Synonyms: para-methoxyphenylpiperazine hydrochloridyl
Molecular Weight (g/mol): 227.71
Molecular Formula: C11H16ClN2O
InChi Key: MFHKWSCQPZNUNF-UHFFFAOYSA-N
SMILES: [Cl].COC1=CC=C(C=C1)N1CCNCC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)