accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3887-48-7

IUPAC Name: hydrogen 3-aminobenzene-1-sulfonyl fluoride chloride
Synonyms: hydrogen 3-aminobenzenesulfonyl fluoride chloride
Molecular Weight (g/mol): 211.64
Molecular Formula: C6H7ClFNO2S
InChi Key: KTADZFWRDLKHKW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC(=C1)S(F)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results