accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 389-58-2

SS

CAS RN 389-58-2

IUPAC Name: 3,11-dithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Synonyms: 3,11-dithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Molecular Weight (g/mol): 178.27
Molecular Formula: C9H6S2
InChi Key: UITASDKJJNYORO-UHFFFAOYSA-N
SMILES: C1C2=C(SC=C2)C2=C1C=CS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results