Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4H-Cyclopenta[2,1-b:3,4-b']dithiophene 97.0+%, TCI America™
SDP

Catalog No. C3021200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3021200MG 200 mg
C30211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3021200MG Supplier TCI America Supplier No. C3021200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 389-58-2
Molecular Formula C9H6S2
Molecular Weight (g/mol) 178.27
MDL Number MFCD20133900
InChI Key UITASDKJJNYORO-UHFFFAOYSA-N
Synonym 4H-Cyclopenta[1,2-b:5,4-b′C]dithiophene
PubChem CID 136232
IUPAC Name 3,11-dithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
SMILES C1C2=C(SC=C2)C2=C1C=CS2

Specifications

Melting Point 73°C
Quantity 200 mg
Formula Weight 178.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4H-Cyclopenta[2,1-b:3,4-b′]dithiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.