accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3893-23-0

IUPAC Name: 2-cyclohexyl-2-phenylacetonitrile
Synonyms: 2-cyclohexyl-2-phenylacetonitrile
Molecular Weight (g/mol): 199.3
Molecular Formula: C14H17N
InChi Key: IZSWBXTYTALSOZ-UHFFFAOYNA-N
SMILES: N#CC(C1CCCCC1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)