Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyclohexylphenylacetonitrile, 98%

Catalog No. AAL0535018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0535018 50 g
AAL0535009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0535018 Supplier Thermo Scientific Chemicals Supplier No. L0535018
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3893-23-0 | C14H17N | 199.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3893-23-0
Molecular Formula C14H17N
Molecular Weight (g/mol) 199.30
MDL Number MFCD00019362
InChI Key IZSWBXTYTALSOZ-UHFFFAOYNA-N
Synonym cyclohexylphenylacetonitrile, cyclohexyl phenyl acetonitrile, alpha-cyclohexylphenylacetonitrile, benzeneacetonitrile, .alpha.-cyclohexyl, 2-cyclohexyl-2-phenylethanenitrile, acmc-20alcf, cyclohexyl-phenylacetonitrile, alpha-phenylcyclohexaneacetonitrile, 2-cyclohexyl-2-phenyl-acetonitrile, .alpha.-cyclohexylphenylacetonitrile
PubChem CID 95302
IUPAC Name 2-cyclohexyl-2-phenylacetonitrile
SMILES N#CC(C1CCCCC1)C1=CC=CC=C1

Specifications

Melting Point 52°C to 56°C
Boiling Point 137°C to 140°C (3 mmHg)
Quantity 50 g
UN Number UN3439
Beilstein 2051115
Formula Weight 199.3
Percent Purity 98%
Chemical Name or Material alpha-Cyclohexylphenylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 223-442-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.