CAS RN 3894-09-5
IUPAC Name:
(2R)-2-cyclohexyl-2-phenylacetate
Synonyms:
(2R)-2-cyclohexyl-2-phenylacetate
Molecular Weight (g/mol):
217.29
Molecular Formula:
C14H17O2
InChi Key:
AAJLPPDFIRPBDA-ZDUSSCGKSA-M
SMILES:
[O-]C(=O)[C@H](C1CCCCC1)C1=CC=CC=C1
1
–
1
of
1
results
1
alpha-Cyclohexylphenylacetic Acid 95.0+%, TCI America™
CAS: 3894-09-5 Molecular Formula: C14H17O2 Molecular Weight (g/mol): 217.29 MDL Number: MFCD00001477 InChI Key: AAJLPPDFIRPBDA-ZDUSSCGKSA-M Synonym: 2-Cyclohexyl-2-phenylacetic Acid PubChem CID: 97942 IUPAC Name: 2-cyclohexyl-2-phenylacetic acid SMILES: C1CCC(CC1)C(C2=CC=CC=C2)C(=O)O