Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyclohexylphenylacetic Acid 95.0+%, TCI America™
SDP

Catalog No. C28125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28125G 5 g
C28121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28125G Supplier TCI America Supplier No. C28125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3894-09-5
Molecular Formula C14H17O2
Molecular Weight (g/mol) 217.29
MDL Number MFCD00001477
InChI Key AAJLPPDFIRPBDA-ZDUSSCGKSA-M
Synonym 2-Cyclohexyl-2-phenylacetic Acid
PubChem CID 97942
IUPAC Name 2-cyclohexyl-2-phenylacetic acid
SMILES C1CCC(CC1)C(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 5 g
Formula Weight 218.30
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Cyclohexylphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.