accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OBOHHO

CAS RN 389621-80-1

IUPAC Name: [4-(diethylcarbamoyl)phenyl]boronic acid
Synonyms: 4-(diethylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 221.06
Molecular Formula: C11H16BNO3
InChi Key: ZCGVBHIMRVYWOH-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results