Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Diethylcarbamoyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D46815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4681-5G 5 g
D46811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4681-5G Supplier TCI America Supplier No. D46815G
Add to Cart
Add to Cart

Specifications

CAS 389621-80-1
Melting Point 142°C
Color White
Molecular Formula C11H16BNO3
Quantity 5 g
MDL Number MFCD03411949
Synonym 4-n,n-diethylaminocarbonyl phenylboronic acid, 4-diethylcarbamoyl phenylboronic acid, 4-diethylcarbamoyl phenyl boronic acid, 4-diethylcarbamoyl benzeneboronic acid, boronic acid, 4-diethylamino carbonyl phenyl
InChI Key ZCGVBHIMRVYWOH-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C(=O)N(CC)CC)(O)O
IUPAC Name [4-(diethylcarbamoyl)phenyl]boronic acid
Molecular Weight (g/mol) 221.063
PubChem CID 2773375
Formula Weight 221.06
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.