accessibility menu, dialog, popup

UserName

Viewing profile as user:
BClHOOH

CAS RN 3900-89-8

IUPAC Name: (2-chlorophenyl)boronic acid
Synonyms: 2-chlorophenylboronic acid
Molecular Weight (g/mol): 156.37
Molecular Formula: C6H6BClO2
InChi Key: RRCMGJCFMJBHQC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (6)
  • (1)
  • (1)
Quantity 
  • (3)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)