Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C17055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1705-5G 5 g
C17051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1705-5G Supplier TCI America Supplier No. C17055G
Add to Cart
Add to Cart

Specifications

CAS 3900-89-8
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C6H6BClO2
MDL Number MFCD00674012
Synonym 2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid
InChI Key RRCMGJCFMJBHQC-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1Cl)(O)O
IUPAC Name (2-chlorophenyl)boronic acid
Molecular Weight (g/mol) 156.372
PubChem CID 2734322
Formula Weight 156.37
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.