accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOI

CAS RN 39028-27-8

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-iodoacetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-iodoacetate
Molecular Weight (g/mol): 283.02
Molecular Formula: C6H6INO4
InChi Key: VRDGQQTWSGDXCU-UHFFFAOYSA-N
SMILES: ICC(=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)