Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl Iodoacetate 95.0+%, TCI America™
SDP

Catalog No. S0844100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0844100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0844100MG Supplier TCI America Supplier No. S0844100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39028-27-8
Molecular Formula C6H6INO4
Molecular Weight (g/mol) 283.021
MDL Number MFCD00058451
InChI Key VRDGQQTWSGDXCU-UHFFFAOYSA-N
Synonym Iodoacetic Acid N-Succinimidyl Ester, N-(Iodoacetoxy)succinimide
PubChem CID 3299230
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-iodoacetate
SMILES C1CC(=O)N(C1=O)OC(=O)CI

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 100 mg
Formula Weight 283.02
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl Iodoacetate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.