accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2NO

CAS RN 39070-63-8

IUPAC Name: 4-benzoylbenzene-1,2-diamine
Synonyms: 4-benzoylbenzene-1,2-diamine
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: RXCOGDYOZQGGMK-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1N)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (4)
Physical Form 
  • (2)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)