accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39093-93-1

IUPAC Name: 1λ⁶-thiomorpholine-1,1-dione
Synonyms: 1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 135.18
Molecular Formula: C4H9NO2S
InChi Key: NDOVLWQBFFJETK-UHFFFAOYSA-N
SMILES: O=S1(=O)CCNCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)