Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiomorpholine 1,1-dioxide, Thermo Scientific Chemicals

Catalog No. AC466030250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466030250 25 g
AC466030050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466030250 Supplier Thermo Scientific Chemicals Supplier No. 466030250
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39093-93-1 | C4H9NO2S | 135.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39093-93-1
Molecular Formula C4H9NO2S
Molecular Weight (g/mol) 135.18
InChI Key NDOVLWQBFFJETK-UHFFFAOYSA-N
IUPAC Name 1λ⁶-thiomorpholine-1,1-dione
SMILES O=S1(=O)CCNCC1

Specifications

Melting Point 68.0°C to 72.0°C
Quantity 25 g
Formula Weight 135.18
Chemical Name or Material Thiomorpholine 1,1-dioxide
Hazard Category GHS Signal Word: Warning
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wear protective gloves/protective clothing/eye protection/face protection.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.