accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39115-96-3

IUPAC Name: 3-bromobenzohydrazide
Synonyms: benzoylhydrazine, M-bromo
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: BNAQRAZIPAHWAR-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC(Br)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)