Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzhydrazide, 98+%

Catalog No. AAA1962814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1962814 25 g
AAA1962806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1962814 Supplier Thermo Scientific Chemicals Supplier No. A1962814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39115-96-3 | C7H7BrN2O | 215.05 g/mol

3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.

Solubility
Slightly soluble in water (3.7 g/L) (25°C)

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39115-96-3
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00007600
InChI Key BNAQRAZIPAHWAR-UHFFFAOYSA-N
Synonym 3-bromobenzhydrazide, m-bromobenzohydrazide, 3-bromobenzoic hydrazide, benzoic acid, 3-bromo-, hydrazide, 3-bromobenzoic acid hydrazide, 3-bromobenzoylhydrazine, benzoylhydrazine, m-bromo, 3-bromo-benzoic acid hydrazide, 3-bromobenzenecarbohydrazide, 3-bromanylbenzohydrazide
PubChem CID 520941
IUPAC Name 3-bromobenzohydrazide
SMILES C1=CC(=CC(=C1)Br)C(=O)NN

Specifications

Melting Point 154°C to 157°C
Quantity 25 g
Beilstein 1945814
Solubility Information Slightly soluble in water (3.7g/L) (25°C)
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 254-298-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.