accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO(Z)

CAS RN 3913-85-7

IUPAC Name: (2Z)-dec-2-enoic acid
Synonyms: cis-2-decenoic acid (Z)-2-decensaeure
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: WXBXVVIUZANZAU-HJWRWDBZSA-N
SMILES: CCCCCCC\C=C/C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results