Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Decenoic Acid, MP Biomedicals

Catalog No. ICN202877 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202877 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202877 Supplier MP Biomedicals, Inc Supplier No. 0520287710

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3913-85-7
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
InChI Key WXBXVVIUZANZAU-HJWRWDBZSA-N
Synonym z-2-decenoic acid, cis-2-decenoic acid, z-dec-2-enoic acid, cis-dec-2-enoic acid, 2z-decenoic acid, 2-cis-decenoic acid, z-2-decensaeure, 2z-dec-2-enoic acid, c10:1, n-8 cis, cis-2-decenoic acid hplc
PubChem CID 5356596
ChEBI CHEBI:50466
IUPAC Name (Z)-dec-2-enoic acid
SMILES CCCCCCCC=CC(=O)O

Specifications

Formula Weight 170
Quantity 10 g
Chemical Name or Material 2-Decenoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.