accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39149-80-9

CH3OOCH3H3CCH3Br

CAS RN 39149-80-9

IUPAC Name: tert-butyl 2-bromopropanoate
Synonyms: tert-butyl 2-bromopropanoate
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: CVAWKJKISIPBOD-UHFFFAOYNA-N
SMILES: CC(Br)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results