Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 2-Bromopropionate 97.0+%, TCI America™
SDP

Catalog No. B1277500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1277500G 500 g
B127725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1277500G Supplier TCI America Supplier No. B1277500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39149-80-9
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00043897
InChI Key CVAWKJKISIPBOD-UHFFFAOYSA-N
Synonym 2-Bromopropionic Acid tert-Butyl Ester
PubChem CID 545327
IUPAC Name tert-butyl 2-bromopropanoate
SMILES CC(C(=O)OC(C)(C)C)Br

Specifications

Color Yellow
Boiling Point 62°C
Quantity 500 g
Formula Weight 209.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl 2-Bromopropionate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.