accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39156-41-7

IUPAC Name: 4-methoxybenzene-1,3-diamine; sulfuric acid
Synonyms: 2,4-diaminoanisole sulfate
Molecular Weight (g/mol): 236.24
Molecular Formula: C7H12N2O5S
InChi Key: FDJQHYWUHGBBTO-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.COC1=CC=C(N)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results