Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-m-phenylenediamine Sulfate Hydrate 98%, Thermo Scientific™

Catalog No. AC179492500
Change view
Click to view available options
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC179492500 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC179492500 Supplier Thermo Scientific Chemicals Supplier No. 179492500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39156-41-7
CAS Min % 97.5
CAS Max % 100.0
Molecular Formula C7H12N2O5S
Linear Formula CH3OC6H3(NH2)2.H2SO4.xH2O
Quantity 250g
Merck Index 15, 2978
Synonym 2,4-diaminoanisole sulfate, 4-methoxybenzene-1,3-diamine sulfate, pelagol grey, fouramine ba, fourrine sla, pelagol ba, pelagol sla, durafur brown mn, furro sla, renal sla
InChI Key FDJQHYWUHGBBTO-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)N)N.OS(=O)(=O)O
IUPAC Name 4-methoxybenzene-1,3-diamine;sulfuric acid
Molecular Weight (g/mol) 236.242
PubChem CID 38221
Formula Weight 236.26
Chemical Name or Material 4-Methoxy-m-phenylenediamine sulfate hydrate, 98%
Show More Show Less
EINECSNumber 254-323-9
RTECSNumber ST2705000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.