accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClFCl

CAS RN 392-71-2

IUPAC Name: 2,6-dichloro-4-fluorophenol
Synonyms: 2,6-dichloro-4-fluorophenol
Molecular Weight (g/mol): 180.99
Molecular Formula: C6H3Cl2FO
InChi Key: BOJVIFKSTRCIRJ-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(F)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)