Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-4-fluorophenol, TCI America™
SDP

Catalog No. D220410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D220410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D220410G Supplier TCI America Supplier No. D220410G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 392-71-2
Molecular Formula C6H3Cl2FO
Molecular Weight (g/mol) 180.987
MDL Number MFCD00010675
InChI Key BOJVIFKSTRCIRJ-UHFFFAOYSA-N
Synonym phenol, 2,6-dichloro-4-fluoro, 2,6-dichloro-4-fluoro-phenol, pubchem2871, acmc-1ai43, 4-fluoro-2,6-dichlorophenol, bojvifkstrcirj-uhfffaoysa, phenol,2,6-dichloro-4-fluoro, attercop-chm at126185
PubChem CID 94950
IUPAC Name 2,6-dichloro-4-fluorophenol
SMILES C1=C(C=C(C(=C1Cl)O)Cl)F

Specifications

Melting Point 49°C
Color White
Quantity 10 g
Formula Weight 180.99
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-4-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.