accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39201-33-7

HOON

CAS RN 39201-33-7

IUPAC Name: 4-cyanobutanoic acid
Synonyms: 4-cyanobutanoic acid
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: YXBVMSQDRLXPQV-UHFFFAOYSA-N
SMILES: OC(=O)CCCC#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results