Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanobutyric acid, 95%

Catalog No. AAH66674MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66674MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66674MD Supplier Thermo Scientific Chemicals Supplier No. H66674MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39201-33-7 | C5H7NO2 | 113.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39201-33-7
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.116
MDL Number MFCD00218781
InChI Key YXBVMSQDRLXPQV-UHFFFAOYSA-N
Synonym 4-cyanobutyric acid, butanoic acid, 4-cyano, 4-cyanobutanoicacid
PubChem CID 38239
IUPAC Name 4-cyanobutanoic acid
SMILES C(CC#N)CC(=O)O

Specifications

Quantity 250 mg
UN Number UN2923
Beilstein 1746747
Formula Weight 113.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Cyanobutyric acid
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.