CAS RN 39213-05-3
IUPAC Name:
2-chloro-4-(morpholin-4-yl)quinazoline
Synonyms:
2-chloro-4-(morpholin-4-yl)quinazoline
Molecular Weight (g/mol):
249.7
Molecular Formula:
C12H12ClN3O
InChi Key:
NJBDGZVAZRMIFM-UHFFFAOYSA-N
SMILES:
ClC1=NC(N2CCOCC2)=C2C=CC=CC2=N1
1
–
1
of
1
results
1
2-Chloro-4-(4-morpholinyl)quinazoline, 97%, Thermo Scientific™
CAS: 39213-05-3 Molecular Formula: C12H12ClN3O Molecular Weight (g/mol): 249.70 MDL Number: MFCD03083869 InChI Key: NJBDGZVAZRMIFM-UHFFFAOYSA-N Synonym: 4-2-chloroquinazolin-4-yl morpholine,2-chloro-4-morpholino-quinazoline,2-chloro-4-morpholin-4-yl quinazoline,2-chloro-4-morpholinoquinazoline,2-chloro-4-4-morpholinyl quinazoline,2-chloro-4-morpholin-4-yl-quinazoline PubChem CID: 2338466 IUPAC Name: 4-(2-chloroquinazolin-4-yl)morpholine SMILES: ClC1=NC(N2CCOCC2)=C2C=CC=CC2=N1