accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39253-43-5

IUPAC Name: N-(4-methoxyphenyl)-4-methylaniline
Synonyms: N-(4-methoxyphenyl)-4-methylaniline
Molecular Weight (g/mol): 213.28
Molecular Formula: C14H15NO
InChi Key: KIDXWDVZFZMXGM-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC2=CC=C(C)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results