Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-4'-methyldiphenylamine 98.0+%, TCI America™
SDP

Catalog No. M20705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20705G 5 g
M207025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20705G Supplier TCI America Supplier No. M20705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39253-43-5
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00092922
InChI Key KIDXWDVZFZMXGM-UHFFFAOYSA-N
Synonym N-(4-Methoxyphenyl)-p-tolylamine
PubChem CID 11435828
IUPAC Name N-(4-methoxyphenyl)-4-methylaniline
SMILES COC1=CC=C(NC2=CC=C(C)C=C2)C=C1

Specifications

Melting Point 84°C
Quantity 5 g
Formula Weight 213.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-4′-methyldiphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.