accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39262-22-1

IUPAC Name: {7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl}methanesulfonyl chloride
Synonyms: {7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl}methanesulfonyl chloride
Molecular Weight (g/mol): 250.74
Molecular Formula: C10H15ClO3S
InChi Key: BGABKEVTHIJBIW-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(CS(Cl)(=O)=O)C(=O)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results