Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-10-Camphorsulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C13085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13085G 5 g
C130825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13085G Supplier TCI America Supplier No. C13085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39262-22-1
Molecular Formula C10H15ClO3S
Molecular Weight (g/mol) 250.737
MDL Number MFCD00064155
InChI Key BGABKEVTHIJBIW-XVKPBYJWSA-N
Synonym 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, l--10-camphorsulfonyl chloride, l---10-camphorsulfonyl chloride, 10-camphorsulfonyl chloride, 1r---camphor-10-sulfonyl chloride, 1r-camphor-10-sulfonic acid chloride, pubchem8092, ksc914s0b, l-10-camphorsulfonyl chloride
PubChem CID 9816396
IUPAC Name [(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C

Specifications

Melting Point 67°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 250.74
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-10-Camphorsulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.