CAS RN 39280-86-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Glycol chitosan, 76.2%, MP Biomedicals™
CAS: 39280-86-9 Molecular Formula: (C8H15NO5)n