accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONF

CAS RN 393-55-5

IUPAC Name: 2-fluoropyridine-3-carboxylic acid
Synonyms: 2-fluoropyridine-3-carboxylic acid
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: LLLVHTWJGWNRBD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CN=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results