Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoronicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. F05755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05755G Supplier TCI America Supplier No. F05755G
Add to Cart
Add to Cart

Specifications

CAS 393-55-5
Melting Point 165°C
Color White-Yellow
Molecular Formula C6H4FNO2
Quantity 5 g
MDL Number MFCD00040744
Synonym 2-fluoronicotinic acid, 2-fluoro-3-pyridinecarboxylic acid, 2-fluoro-nicotinic acid, 2-fluoro-3-carboxypyridine, 2-fluoronicotinicacid, 3-pyridinecarboxylic acid, 2-fluoro, 2-fluoro-3-pyridinecarboxylic acid 2-fluoronicotinic acid, fluoronicotinic acid, pubchem1120, 2-fluoro nicotinic acid
InChI Key LLLVHTWJGWNRBD-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)F)C(=O)O
IUPAC Name 2-fluoropyridine-3-carboxylic acid
Molecular Weight (g/mol) 141.101
PubChem CID 96089
Formula Weight 141.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoronicotinic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.