accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOH3CCH3XCH3NOXO(E)

CAS RN 39300-45-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 366.41
Molecular Formula: C18H26N2O6
InChi Key: —
SMILES: [O-][N+](*)=O.CCCCCCC(C)*.C\C=C\C(=O)OC1=C(C=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results