accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNClClFFF

CAS RN 3932-97-6

IUPAC Name: 2,4-dichloro-5-(trifluoromethyl)pyrimidine
Synonyms: 2,4-dichloro-5-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 216.97
Molecular Formula: C5HCl2F3N2
InChi Key: IDRUEHMBFUJKAK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(Cl)N=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)