Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific Chemicals

Catalog No. AC454780010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454780010 1 g
AC454780050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454780010 Supplier Thermo Scientific Chemicals Supplier No. 454780010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3932-97-6
Molecular Formula C5HCl2F3N2
Molecular Weight (g/mol) 216.97
MDL Number MFCD03426408
InChI Key IDRUEHMBFUJKAK-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-trifluoromethyl pyrimidine, 2,4-dichloro-5-trifluoromethyl-pyrimidine, 5-trifluoromethyl-2,4-dichloropyrimidine, pyrimidine, 2,4-dichloro-5-trifluoromethyl, pubchem5304, acmc-1cn5t, c5hcl12f3n2, 2,4-dichloro-5-trifluoromethylpyrimidin, 2,6-dichloro-5-trifluoromethylpyrimidine, 2.4-dichloro-5-trifluoromethylpyrimidine
PubChem CID 2782774
IUPAC Name 2,4-dichloro-5-(trifluoromethyl)pyrimidine
SMILES FC(F)(F)C1=CN=C(Cl)N=C1Cl

Specifications

Density 1.6090g/mL
Boiling Point 48.0°C to 50.0°C (32.0 mmHg)
Flash Point 93°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.609
Formula Weight 216.98
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-(trifluoromethyl)pyrimidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYE

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.