accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOOFFF

CAS RN 394-25-2

IUPAC Name: 1-methoxy-2-nitro-4-(trifluoromethyl)benzene
Synonyms: 1-methoxy-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 221.14
Molecular Formula: C8H6F3NO3
InChi Key: MAAFHLOZHBKYTG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results