Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-3-nitrobenzotrifluoride, TCI America™
SDP

Catalog No. T10425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10425G Supplier TCI America Supplier No. T10425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 394-25-2
Molecular Formula C8H6F3NO3
Molecular Weight (g/mol) 221.135
MDL Number MFCD00007099
InChI Key MAAFHLOZHBKYTG-UHFFFAOYSA-N
Synonym 4-methoxy-3-nitrobenzotrifluoride, 1-methoxy-2-nitro-4-trifluoromethyl benzene, 2-nitro-4-trifluoromethyl anisole, 2-nitro-4-trifluoromethylanisole, 3-nitro-4-methoxybenzotrifluoride, 4-trifluoromethyl-2-nitroanisole, benzene, 1-methoxy-2-nitro-4-trifluoromethyl, 4-trifluoromethyl-1-methoxy-2-nitrobenzene, pubchem13493, 4-methoxy-3-nitrobezotrifluoride
PubChem CID 96748
IUPAC Name 1-methoxy-2-nitro-4-(trifluoromethyl)benzene
SMILES COC1=C(C=C(C=C1)C(F)(F)F)[N+](=O)[O-]

Specifications

Melting Point 48°C
Color Yellow
Quantity 5 g
UN Number 1759
Formula Weight 221.14
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3-nitrobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.