CAS RN 39422-83-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Carboxymethyldextran Sodium Salt (Mw.=ca. 40,000), TCI America™
CAS: 39422-83-8 MDL Number: MFCD00146477
2
Carboxymethyldextran Sodium Salt (Mw.=ca. 10,000), TCI America™
CAS: 39422-83-8 MDL Number: MFCD00146477