Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxymethyldextran Sodium Salt (Mw.=ca. 10,000), TCI America™
SDP

Catalog No. C32501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C32501G Supplier TCI America Supplier No. C32501G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39422-83-8
MDL Number MFCD00146477

Specifications

Color White-Yellow
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Carboxymethyldextran Sodium Salt (Mw.=ca. 10,000)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.