accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39430-51-8

IUPAC Name: chromium(3+) triacetate
Synonyms: chromium acetate hydroxide
Molecular Weight (g/mol): 229.13
Molecular Formula: C6H9CrO6
InChi Key: WYYQVWLEPYFFLP-UHFFFAOYSA-K
SMILES: [Cr+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)