Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) acetate hydroxide, Cr 24%

Catalog No. AA12518A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12518A3 2 kg
AA1251836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA12518A3 Supplier Thermo Scientific Chemicals Supplier No. 012518A3
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39430-51-8 | Cr3(OH)2(OOCCH3)7

Occasionally to dehalogenate organic compounds. used as an oxygen scrubber, polymer industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39430-51-8
Molecular Formula Cr3(OH)2(OOCCH3)7
MDL Number MFCD00058912

Specifications

Color Blue
Physical Form Powder
Quantity 2 kg
Odor Odorless
Solubility Information Soluble in water. Insoluble in acetone. Soluble in hot water
Formula Weight 603.31
Chemical Name or Material Chromium(III) acetate hydroxide, Cr 24%
Hazard Category H317
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 254-447-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.