accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39512-51-1

IUPAC Name: 1-(2-methylphenyl)piperazine
Synonyms: 1-(2-methylphenyl)piperazine
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: WICKLEOONJPMEQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1N1CCNCC1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)