Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(o-Tolyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. T34045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34045G 5 g
T340425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34045G Supplier TCI America Supplier No. T34045G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39512-51-1
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD00040729
InChI Key WICKLEOONJPMEQ-UHFFFAOYSA-N
Synonym 1-2-methylphenyl piperazine, 1-o-tolyl piperazine, 4-2-methylphenyl piperazine, 1-o-tolylpiperazine, piperazine, 1-2-methylphenyl, 1-o-tolyl-piperazine, 1-2-methylphenyl-piperazine, o-tolylpiperazine, 2-methylphenylpiperazine, chembl9746
PubChem CID 91965
IUPAC Name 1-(2-methylphenyl)piperazine
SMILES CC1=CC=CC=C1N2CCNCC2

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 161°C
Quantity 5 g
UN Number 3263
Formula Weight 176.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(o-Tolyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.