accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 39515-51-0

IUPAC Name: 3-phenoxybenzaldehyde
Synonyms: 3-phenoxybenzaldehyde
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: MRLGCTNJRREZHZ-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC(OC2=CC=CC=C2)=C1
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)